Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnnt3
Entrez Gene ID
21957
Gene Name
troponin T3, skeletal, fast
Gene Aliases
fTnT
Chromosome Location
Chr.7: 142498777 - 142516213 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163664.1 NP_001157136.1 1 - 2 65 69
NM_001163665.1 NP_001157137.1 1 - 2 65 69
NM_001163666.1 NP_001157138.1 1 - 2 65 69
NM_001163667.1 NP_001157139.1 1 - 2 65 69
NM_001163668.1 NP_001157140.1 1 - 2 65 69
NM_001163669.1 NP_001157141.1 1 - 2 65 69
NM_001163670.1 NP_001157142.1 1 - 2 65 69
NM_011620.3 NP_035750.2 1 - 2 65 69
GenBank mRNA AA028290.1 - 1 - 2 45 69
AA754928.1 - 1 - 2 42 69
AA771596.1 - 1 - 2 43 69
AI173874.1 - 1 - 2 57 69
AI746973.1 - 1 - 2 60 69
BC003747.1 - 1 - 2 61 69
BY766323.1 - 1 - 2 55 69
L49466.1 - 1 - 2 65 69
L49467.1 - 1 - 2 65 69
L49468.1 - 1 - 2 65 69
L49469.1 - 1 - 2 65 69
L49470.1 - 1 - 2 65 69
L49471.1 - 1 - 2 65 69
L49472.1 - 1 - 2 65 69
U77779.1 - 1 - 2 58 69

View Full Product Details