Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St6gal2
Entrez Gene ID
240119
Gene Name
beta galactoside alpha 2,6 sialyltransferase 2
Gene Aliases
C230064G14Rik, St6galII, mKIAA1877
Chromosome Location
Chr.17: 55444358 - 55514581 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172829.2 NP_766417.1 2 - 3 1079 147
GenBank mRNA AK082566.1 - 2 - 3 1080 147
BC125586.1 - 1 - 2 949 147
BC138142.1 - 1 - 2 949 147

View Full Product Details