Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Satb1
Entrez Gene ID
20230
Gene Name
special AT-rich sequence binding protein 1
Gene Aliases
2610306G12Rik, AW413156
Chromosome Location
Chr.17: 51736187 - 51833497 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163630.1 NP_001157102.1 4 - 5 834 70
NM_001163631.1 NP_001157103.1 4 - 5 702 70
NM_001163632.1 NP_001157104.1 4 - 5 675 70
NM_009122.2 NP_033148.2 4 - 5 725 70
GenBank mRNA AK037740.1 - 4 - 5 725 70
AK088563.1 - 4 - 5 701 70
AK134040.1 - 4 - 5 705 70
AK158518.1 - 4 - 5 835 70
BC011132.1 - 4 - 5 585 70
CT010371.1 - 3 - 4 513 70
U05252.1 - 4 - 5 711 70

View Full Product Details