The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164477.1 | NP_001157949.1 | 14 - 15 | 3079 | 138 |
NM_027835.3 | NP_082111.2 | 15 - 16 | 3226 | 138 | |
GenBank mRNA | AF374384.1 | - | 15 - 16 | 3135 | 138 |
AK018602.1 | - | 4 - 5 | 560 | 138 | |
AK153018.1 | - | 15 - 16 | 3133 | 138 | |
AK153344.1 | - | 7 - 8 | 1160 | 138 | |
AY075132.1 | - | 15 - 16 | 2901 | 138 | |
BC004031.1 | - | 8 - 9 | 1368 | 138 | |
BC019605.1 | - | 2 - 3 | 257 | 138 | |
BC025508.1 | - | 10 - 11 | 1670 | 138 | |
BC080200.1 | - | 14 - 15 | 2984 | 138 | |
EF113547.1 | - | 15 - 16 | 2915 | 138 | |
EF113548.1 | - | 15 - 16 | 2915 | 138 | |
EU084750.1 | - | 15 - 16 | 2915 | 138 |