Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fchsd2
Entrez Gene ID
207278
Gene Name
FCH and double SH3 domains 2
Gene Aliases
BC034086, R74866, Sh3md3, mKIAA0769
Chromosome Location
Chr.7: 101108711 - 101284405 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146010.1 NP_001139482.1 16 - 17 1947 84
NM_199012.2 NP_950177.2 17 - 18 2019 84
GenBank mRNA AK122368.1 - 17 - 18 1911 84
AK140322.1 - 16 - 17 1926 84
AK141949.1 - 16 - 17 1947 84
BC034086.1 - 4 - 5 493 84
BC039788.1 - 3 - 4 421 84
BC060647.1 - 17 - 18 1981 84

View Full Product Details