Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zmiz1
Entrez Gene ID
328365
Gene Name
zinc finger, MIZ-type containing 1
Gene Aliases
BC065120, E330020C23, ENSMUSG00000072684, Gm10397, I920194n01, Rai17, Zimp10
Chromosome Location
Chr.14: 25458865 - 25666747 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183208.3 NP_899031.2 8 - 9 1150 126
GenBank mRNA AK054366.1 - 8 - 9 1044 126
BC058646.1 - 8 - 9 1068 126
BC065120.1 - 8 - 9 1043 126

View Full Product Details