Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxp4
Entrez Gene ID
74123
Gene Name
forkhead box P4
Gene Aliases
1200010K03Rik, 2310007G05Rik, mFKHLA
Chromosome Location
Chr.17: 47867133 - 47924645 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110824.1 NP_001104294.1 8 - 9 1443 63
NM_001110825.1 NP_001104295.1 8 - 9 1443 63
NM_028767.2 NP_083043.2 8 - 9 1440 63
GenBank mRNA AB052766.1 - 8 - 9 1274 63
AK004693.1 - 8 - 9 1251 63
AK143194.1 - 8 - 9 1290 63
AY135029.1 - 8 - 9 1231 63
BC043702.1 - 8 - 9 1217 63
BC052407.1 - 8 - 9 1246 63
BC057110.1 - 8 - 9 1440 63

View Full Product Details