The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172673.3 | NP_766261.2 | 4 - 5 | 562 | 69 |
GenBank mRNA | AK030883.1 | - | 4 - 5 | 318 | 69 |
AK044270.1 | - | 4 - 5 | 313 | 69 | |
AK048865.1 | - | 4 - 5 | 563 | 69 | |
AK054484.1 | - | 4 - 5 | 519 | 69 | |
AK146383.1 | - | 4 - 5 | 530 | 69 | |
BC038623.1 | - | 4 - 5 | 299 | 69 | |
BC057549.1 | - | 1 - 2 | 91 | 69 | |
BC062889.1 | - | 4 - 5 | 519 | 69 |