Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
H2-M1
Entrez Gene ID
224756
Gene Name
histocompatibility 2, M region locus 1
Gene Aliases
H-2M1, H2-Qa-Mb1, Mb1
Chromosome Location
Chr.17: 36669030 - 36672197 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177636.2 NP_808304.2 2 - 3 342 165
GenBank mRNA AF539594.1 - 2 - 3 342 165

View Full Product Details