The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168273.1 | NP_001161745.1 | 1 - 2 | 66 | 82 |
NM_172596.2 | NP_766184.2 | 1 - 2 | 66 | 82 | |
GenBank mRNA | AK036519.1 | - | 1 - 2 | 66 | 82 |
AK122199.1 | - | 1 - 2 | 44 | 82 | |
BC040370.1 | - | 1 - 2 | 44 | 82 |