The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145920.2 | NP_001139392.1 | 1 - 2 | 1422 | 74 |
NM_001146038.2 | NP_001139510.1 | 3 - 4 | 687 | 74 | |
NM_001271627.1 | NP_001258556.1 | 2 - 3 | 851 | 74 | |
NM_001271630.1 | NP_001258559.1 | 1 - 2 | 950 | 74 | |
NM_001271631.1 | NP_001258560.1 | 1 - 2 | 950 | 74 | |
NM_009820.5 | NP_033950.2 | 3 - 4 | 654 | 74 | |
NR_073425.1 | 1 - 2 | 950 | 74 | ||
GenBank mRNA | AF005936.1 | - | 2 - 3 | 805 | 74 |
AF010284.1 | - | 3 - 4 | 654 | 74 | |
AF053956.1 | - | 1 - 2 | 140 | 74 | |
AK089291.1 | - | 1 - 2 | 484 | 74 | |
AK134925.1 | - | 1 - 2 | 208 | 74 | |
AK158925.1 | - | 1 - 2 | 950 | 74 | |
D14636.1 | - | 1 - 2 | 1420 | 74 | |
D14637.1 | - | 1 - 2 | 1420 | 74 |