Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vcl
Entrez Gene ID
22330
Gene Name
vinculin
Gene Aliases
9430097D22, AA571387, AI462105, AW545629
Chromosome Location
Chr.14: 20929378 - 21033676 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009502.4 NP_033528.3 10 - 11 1441 88
GenBank mRNA AK077850.1 - 10 - 11 1477 88
BC008520.1 - 10 - 11 1441 88

View Full Product Details