Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Micalcl
Entrez Gene ID
100504195
Gene Name
MICAL C-terminal like
Gene Aliases
4921517J23Rik
Chromosome Location
Chr.7: 112368308 - 112413104 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027587.2 NP_081863.2 4 - 5 1656 83
GenBank mRNA AB359922.1 - 4 - 5 1656 83
AK014911.1 - 1 - 2 1091 83
BC031386.1 - 4 - 5 1830 83

View Full Product Details