Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aars2
Entrez Gene ID
224805
Gene Name
alanyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
Aarsl, AlaRS, Gm89
Chromosome Location
Chr.17: 45506806 - 45520843 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198608.2 NP_941010.2 17 - 18 2367 78
GenBank mRNA AK146958.1 - 17 - 18 2368 78
AK167771.1 - 17 - 18 2362 78
AK173139.1 - 13 - 14 1641 78
AK178778.1 - 2 - 3 206 78
BC059889.1 - 8 - 9 1396 78
BC079844.1 - 17 - 18 2339 78
BC110699.1 - 8 - 9 1396 78
BC113172.1 - 17 - 18 2363 78
BC113779.1 - 17 - 18 2363 78

View Full Product Details