The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081238.2 | NP_001074707.1 | 12 - 13 | 3287 | 75 |
NM_001289844.1 | NP_001276773.1 | 12 - 13 | 2198 | 75 | |
NM_001289845.1 | NP_001276774.1 | 9 - 10 | 1127 | 75 | |
GenBank mRNA | AK156701.1 | - | 12 - 13 | 3206 | 75 |
AK157382.1 | - | 12 - 13 | 3206 | 75 | |
BC044748.2 | - | 8 - 9 | 892 | 75 | |
BC055302.1 | - | 9 - 10 | 1090 | 75 | |
BC066140.1 | - | 13 - 14 | 2096 | 75 |