Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mettl10
Entrez Gene ID
72096
Gene Name
methyltransferase like 10
Gene Aliases
2010208K18Rik
Chromosome Location
Chr.7: 132827232 - 132852880 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028095.1 NP_082371.1 4 - 5 442 158
GenBank mRNA AK008476.1 - 4 - 5 427 158
BC038323.1 - 4 - 5 397 158
BC096622.1 - 4 - 5 426 158
BC116375.2 - 4 - 5 440 158
BQ830246.1 - 4 - 5 468 159

View Full Product Details