Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mettl10
Entrez Gene ID
72096
Gene Name
methyltransferase like 10
Gene Aliases
2010208K18Rik
Chromosome Location
Chr.7: 132827232 - 132852880 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028095.1 NP_082371.1 5 - 6 650 65
GenBank mRNA AK008476.1 - 5 - 6 635 65
BC038323.1 - 5 - 6 605 65
BC096622.1 - 5 - 6 634 65
BC116375.2 - 5 - 6 648 65

View Full Product Details