The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146119.1 | NP_001139591.1 | 3 - 4 | 323 | 86 |
NM_029758.4 | NP_084034.2 | 4 - 5 | 396 | 86 | |
GenBank mRNA | AK047142.1 | - | 3 - 4 | 286 | 86 |
BC049664.1 | - | 4 - 5 | 358 | 86 | |
BC052465.1 | - | 3 - 4 | 304 | 86 |