Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scn2a1
Entrez Gene ID
110876
Gene Name
sodium channel, voltage-gated, type II, alpha 1
Gene Aliases
6430408L10, A230052E19Rik, Nav1.2, Scn2a
Chromosome Location
Chr.2: 65620764 - 65767449 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099298.2 NP_001092768.1 22 - 23 4360 122

View Full Product Details