Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fnbp4
Entrez Gene ID
55935
Gene Name
formin binding protein 4
Gene Aliases
FBP30, Fnbp30, mKIAA1014
Chromosome Location
Chr.2: 90745370 - 90781021 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018828.2 NP_061298.1 1 - 2 460 78
GenBank mRNA AK012167.1 - 1 - 2 273 78
AK014018.1 - 1 - 2 277 78
AK084018.1 - 1 - 2 252 78
AK129263.1 - 1 - 2 357 78
AK164543.1 - 1 - 2 231 78
U40750.2 - 1 - 2 460 78

View Full Product Details