The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038660.2 | NP_001033749.1 | 2 - 3 | 913 | 101 |
NM_177802.3 | NP_808470.1 | 3 - 4 | 470 | 101 | |
GenBank mRNA | AK033481.1 | - | 3 - 4 | 470 | 101 |
AY795939.1 | - | 2 - 3 | 892 | 101 |