Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nol10
Entrez Gene ID
217431
Gene Name
nucleolar protein 10
Gene Aliases
Gm67
Chromosome Location
Chr.12: 17348493 - 17430095 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008421.1 NP_001008421.1 11 - 12 969 88
GenBank mRNA AK148210.1 - 11 - 12 985 88
AK160864.1 - 11 - 12 1005 88
AK164400.1 - 11 - 12 995 88
AK166160.1 - 11 - 12 998 88
BC086676.1 - 11 - 12 969 88

View Full Product Details