Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rag1
Entrez Gene ID
19373
Gene Name
recombination activating gene 1
Gene Aliases
Rag-1
Chromosome Location
Chr.2: 101638252 - 101649532 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009019.2 NP_033045.2 1 - 2 106 74
GenBank mRNA AK088725.1 - 1 - 2 107 74
AK162974.1 - 1 - 2 106 74
M29475.1 - 1 - 2 75 74

View Full Product Details