The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175034.3 | NP_778199.2 | 3 - 4 | 438 | 83 |
GenBank mRNA | AK089225.1 | - | 3 - 4 | 442 | 83 |
AY236527.1 | - | 3 - 4 | 408 | 83 | |
BC094232.1 | - | 3 - 4 | 398 | 83 | |
BC144866.1 | - | 3 - 4 | 433 | 83 | |
BC145643.1 | - | 3 - 4 | 433 | 83 |