Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppp1r10
Entrez Gene ID
52040
Gene Name
protein phosphatase 1, regulatory subunit 10
Gene Aliases
2610025H06Rik, Cat53, D17Ertd808e, Fb19, Pnuts
Chromosome Location
Chr.17: 35916512 - 35952996 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163818.1 NP_001157290.1 17 - 18 2492 71
GenBank mRNA AJ504718.1 - 17 - 18 2498 71
AK145485.1 - 16 - 17 1936 71
BC029765.1 - 9 - 10 1166 71
BC052059.1 - 17 - 18 2495 71

View Full Product Details