Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp1r10
Entrez Gene ID
52040
Gene Name
protein phosphatase 1, regulatory subunit 10
Gene Aliases
2610025H06Rik, Cat53, D17Ertd808e, Fb19, Pnuts
Chromosome Location
Chr.17: 35916512 - 35952996 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163818.1 NP_001157290.1 1 - 2 526 81
GenBank mRNA AJ504718.1 - 1 - 2 532 81
AK053183.1 - 2 - 3 545 81
BC030903.1 - 1 - 2 533 81
BC052059.1 - 1 - 2 529 81

View Full Product Details