Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ebf4
Entrez Gene ID
228598
Gene Name
early B cell factor 4
Gene Aliases
Ebf3, O/E-4, Olf-1
Chromosome Location
Chr.2: 130295249 - 130370481 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110513.1 NP_001103983.1 13 - 14 1419 139
GenBank mRNA AF387630.1 - 12 - 13 1514 139
AF387631.1 - 13 - 14 1419 139
AF387632.1 - 13 - 14 1419 139
AF387633.1 - 13 - 14 1419 139
AF387634.1 - 13 - 14 1419 139
AK149248.1 - 2 - 3 441 139
BC138119.1 - 13 - 14 1419 139
BC145155.1 - 14 - 15 1505 139

View Full Product Details