The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207105.3 | NP_996988.2 | 5 - 6 | 873 | 126 |
GenBank mRNA | AK077436.1 | - | 5 - 6 | 861 | 126 |
AK170370.1 | - | 5 - 6 | 855 | 126 | |
CK330363.1 | - | 1 - 2 | 126 |