Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mcmbp
Entrez Gene ID
210711
Gene Name
MCM (minichromosome maintenance deficient) binding protein
Gene Aliases
1110007A13Rik, AU017382, BC025641, MCM-BP
Chromosome Location
Chr.7: 128696441 - 128740429 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145955.3 NP_666067.1 10 - 11 1362 114
GenBank mRNA AK083385.1 - 10 - 11 1423 114
AK189603.1 - 2 - 3 224 114
BC025641.1 - 10 - 11 1392 114
BC038342.1 - 10 - 11 1362 114

View Full Product Details