Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mcmbp
Entrez Gene ID
210711
Gene Name
MCM (minichromosome maintenance deficient) binding protein
Gene Aliases
1110007A13Rik, AU017382, BC025641, MCM-BP
Chromosome Location
Chr.7: 128696441 - 128740429 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145955.3 NP_666067.1 3 - 4 522 130
GenBank mRNA AK083385.1 - 3 - 4 582 130
BC025641.1 - 3 - 4 552 130
BC038342.1 - 3 - 4 522 130

View Full Product Details