Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sestd1
Entrez Gene ID
228071
Gene Name
SEC14 and spectrin domains 1
Gene Aliases
1500031J16Rik, A930010O20, Solo/Trio8
Chromosome Location
Chr.2: 77180340 - 77280592 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175465.6 NP_780674.1 16 - 17 1888 63
GenBank mRNA AK166483.1 - 3 - 4 345 63
BC011970.1 - 6 - 7 968 63
BC013245.1 - 17 - 18 1969 63
BC038005.1 - 3 - 4 399 63
BC044853.1 - 16 - 17 1888 63
BC048358.1 - 4 - 5 441 63
CN664477.1 - 2 - 3 293 63

View Full Product Details