Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Btbd10
Entrez Gene ID
68815
Gene Name
BTB (POZ) domain containing 10
Gene Aliases
1110056N09Rik, Gmrp1
Chromosome Location
Chr.7: 113315631 - 113370531 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133700.2 NP_598461.1 1 - 2 140 108
GenBank mRNA BC050781.1 - 1 - 2 140 108

View Full Product Details