Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdsub1
Entrez Gene ID
72137
Gene Name
WD repeat, SAM and U-box domain containing 1
Gene Aliases
1700048E19Rik, 2610014F08Rik
Chromosome Location
Chr.2: 59852364 - 59882625 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159636.1 NP_001153108.1 7 - 8 1077 66
NM_028118.2 NP_082394.1 7 - 8 1077 66
GenBank mRNA AK011391.1 - 7 - 8 1013 66
BC050792.1 - 7 - 8 1018 66
BC061948.1 - 7 - 8 1055 66

View Full Product Details