Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnf24
Entrez Gene ID
51902
Gene Name
ring finger protein 24
Gene Aliases
2810473M14Rik, 4930505A13Rik, AI317164, C86507, D2Ertd504e
Chromosome Location
Chr.2: 131298488 - 131352892 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK015699.1 - 5 - 6 457 85

View Full Product Details