The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290656.1 | NP_001277585.1 | 9 - 10 | 4618 | 94 |
NM_029924.2 | NP_084200.1 | 16 - 17 | 6372 | 94 | |
GenBank mRNA | AK173176.1 | - | 9 - 10 | 5239 | 94 |
BC050251.1 | - | 5 - 6 | 3130 | 94 | |
BC138375.1 | - | 9 - 10 | 4523 | 94 | |
BC138378.1 | - | 9 - 10 | 4523 | 94 | |
BU610422.1 | - | 3 - 4 | 467 | 94 |