The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013826.2 | NP_001013848.1 | 3 - 4 | 517 | 68 |
GenBank mRNA | AK075665.1 | - | 3 - 4 | 518 | 68 |
BC147517.1 | - | 3 - 4 | 480 | 68 | |
BC147531.1 | - | 3 - 4 | 480 | 68 |