Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mllt4
- Entrez Gene ID
-
17356
- Gene Name
- myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 4
- Gene Aliases
-
5033403D15Rik, AF6, Af-6, Afadin, Gm314, I-afadin, S-afadin
- Chromosome Location
-
Chr.17: 13760541 - 13905798 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_010806.1 |
NP_034936.1 |
22 - 23 |
3209 |
68 |
GenBank mRNA |
CN538841.1 |
- |
4 - 5 |
430 |
68 |
View Full Product Details