The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010806.1 | NP_034936.1 | 32 - 33 | 5448 | 60 |
GenBank mRNA | BC026648.1 | - | 2 - 3 | 726 | 60 |
BI991173.1 | - | 2 - 3 | 287 | 60 |