Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mllt4
Entrez Gene ID
17356
Gene Name
myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 4
Gene Aliases
5033403D15Rik, AF6, Af-6, Afadin, Gm314, I-afadin, S-afadin
Chromosome Location
Chr.17: 13760541 - 13905798 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010806.1 NP_034936.1 5 - 6 878 104
GenBank mRNA AF172447.1 - 5 - 6 877 104
AK016557.1 - 5 - 6 1042 104

View Full Product Details