Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gen1
Entrez Gene ID
209334
Gene Name
Gen homolog 1, endonuclease (Drosophila)
Gene Aliases
5830483C08Rik
Chromosome Location
Chr.12: 11239718 - 11266029 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177331.4 NP_796305.3 13 - 14 1626 95
GenBank mRNA AK030995.1 - 13 - 14 1636 95
AK031077.1 - 13 - 14 1605 95
AK158255.1 - 12 - 13 1449 95
BC090653.1 - 13 - 14 1626 95
BC099936.1 - 11 - 12 1373 95
BC138157.1 - 13 - 14 1513 95
BC138158.1 - 13 - 14 1513 95

View Full Product Details