Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tcte2
- Entrez Gene ID
-
21646
- Gene Name
- t-complex-associated testis expressed 2
- Gene Aliases
-
AI429268, Tcte-2
- Chromosome Location
-
Chr.17: 13716436 - 13761394 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK005879.1 |
- |
- |
192 |
91 |
View Full Product Details