Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uvrag
Entrez Gene ID
78610
Gene Name
UV radiation resistance associated gene
Gene Aliases
9530039D02Rik, AI648770, BB124205, Uvrag1, Uvragl
Chromosome Location
Chr.7: 98886742 - 99141168 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178635.3 NP_848750.3 8 - 9 1019 77
GenBank mRNA AB020334.1 - 7 - 8 624 77
AK030814.1 - 8 - 9 950 77
AK043084.1 - 8 - 9 997 77
AK044129.1 - 9 - 10 1115 77
AK078085.1 - 8 - 9 1011 77
AK089900.1 - 8 - 9 1004 77
AK154903.1 - 8 - 9 1019 77
AK165015.1 - 8 - 9 1009 77
AK165185.1 - 8 - 9 999 77
BC034072.1 - 2 - 3 205 77
BC034176.1 - 8 - 9 988 77

View Full Product Details