Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Enpp5
Entrez Gene ID
83965
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 5
Gene Aliases
AW061004, D17Abb1e, E-NPP 5, mKIAA4152
Chromosome Location
Chr.17: 44078796 - 44086567 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168620.1 NP_001162091.1 1 - 2 49 74

View Full Product Details