Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vsx1
Entrez Gene ID
114889
Gene Name
visual system homeobox 1 homolog (zebrafish)
Gene Aliases
CHX10-like
Chromosome Location
Chr.2: 150680702 - 150689137 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054068.2 NP_473409.1 1 - 2 620 143
GenBank mRNA AF391758.1 - 1 - 2 465 143
AF395732.1 - 1 - 2 572 143
AK044393.1 - 1 - 2 502 143
AK044687.1 - 1 - 2 502 143
AK157311.1 - 1 - 2 843 143
AY056825.1 - 1 - 2 485 143
BC057647.1 - 1 - 2 620 143
BC138456.1 - 1 - 2 550 143
BC138460.1 - 1 - 2 550 143

View Full Product Details