Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ninl
Entrez Gene ID
78177
Gene Name
ninein-like
Gene Aliases
4930519N13Rik, Gm1004, Gm1634, mKIAA0980
Chromosome Location
Chr.2: 150934519 - 151009398 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207204.2 NP_997087.2 15 - 16 2093 114
GenBank mRNA BC067071.1 - 15 - 16 2027 114

View Full Product Details