Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mocs1
Entrez Gene ID
56738
Gene Name
molybdenum cofactor synthesis 1
Gene Aliases
3110045D15Rik, AI043170, AW536397
Chromosome Location
Chr.17: 49428364 - 49455435 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020042.2 NP_064426.2 6 - 7 837 77
NM_028464.1 NP_082740.1 6 - 7 837 77
GenBank mRNA AK157231.1 - 6 - 7 793 77
BC034066.1 - 2 - 3 186 77
BC049914.1 - 4 - 5 465 77

View Full Product Details