Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mocs1
Entrez Gene ID
56738
Gene Name
molybdenum cofactor synthesis 1
Gene Aliases
3110045D15Rik, AI043170, AW536397
Chromosome Location
Chr.17: 49428364 - 49455435 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020042.2 NP_064426.2 7 - 8 948 84
NM_028464.1 NP_082740.1 7 - 8 948 84
GenBank mRNA AK157231.1 - 7 - 8 904 84
BC034066.1 - 3 - 4 297 84
BC049914.1 - 5 - 6 576 84

View Full Product Details