Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Spats1
- Entrez Gene ID
-
71020
- Gene Name
- spermatogenesis associated, serine-rich 1
- Gene Aliases
-
1700011H05Rik, 4933400B06Rik, Daip2, Ddip, Srsp1
- Chromosome Location
-
Chr.17: 45445877 - 45475194 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK016586.1 |
- |
5 - 6 |
657 |
91 |
AK078278.1 |
- |
- |
652 |
91 |
View Full Product Details