The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146248.2 | NP_666360.2 | 7 - 8 | 1114 | 64 |
GenBank mRNA | AK159780.1 | - | 7 - 8 | 1041 | 64 |
BC031416.1 | - | 8 - 9 | 1203 | 64 | |
CD562711.1 | - | 1 - 2 | 85 | 64 |