The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025408.2 | NP_079684.2 | 10 - 11 | 829 | 124 |
GenBank mRNA | AK004287.1 | - | 10 - 11 | 830 | 124 |
AK011668.1 | - | 10 - 11 | 810 | 124 | |
AK080977.1 | - | 10 - 11 | 807 | 124 | |
AK150629.1 | - | 10 - 11 | 790 | 124 | |
AK162403.1 | - | 10 - 11 | 845 | 124 | |
BC023924.1 | - | 11 - 12 | 997 | 124 |